Gene Transcripts

Gene Symbol
Ogn
Entrez Gene ID
18295
Gene Name
osteoglycin
Gene Aliases
3110079A16Rik, OG, OIF, SLRR3A, mimecan, mimican
Chromosome Location
Chr. 13: 49608071 - 49624500 on Build GRCm38
UniGene ID
Mm.4258
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008760.4 NP_032786.1 2 256
GenBank mRNA AK028300.1 - 2 282
AK053650.1 - 2 257
AK075638.1 - 1 213
AK160760.1 - 2 278
AK163772.1 - 1 209
BC021939.1 - 1 159 5067073
D31951.1 - 1 169

View Full Product Details