Gene Transcripts

Gene Symbol
Cdk18
Entrez Gene ID
18557
Gene Name
cyclin-dependent kinase 18
Gene Aliases
AA682070, Ctk18, Pctk3
Chromosome Location
Chr. 1: 132113547 - 132139957 on Build GRCm38
UniGene ID
Mm.28130
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008795.2 NP_032821.1 16 2067
GenBank mRNA AK004998.1 - 16 2011
AK171569.1 - 16 1849
X69026.1 - 16 2084

View Full Product Details