Gene Transcripts

Gene Symbol
Kcnab1
Entrez Gene ID
16497
Gene Name
potassium voltage-gated channel, shaker-related subfamily, beta member 1
Gene Aliases
Akr8a8, Kvbeta1.1, mKv(beta)1
Chromosome Location
Chr. 3: 64949049 - 65378225 on Build GRCm38
UniGene ID
Mm.316402
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289450.1 NP_001276379.1 15 2680
NM_010597.4 NP_034727.3 14 3096
GenBank mRNA AK004666.1 - 15 2664
AK153200.1 - 15 2681
BC014701.1 - 14 2821 4482268
BY515823.1 - 1 273
X97281.1 - 14 3080

View Full Product Details