Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004869.3 | NP_004860.2 | 11 | 1867 | |
XM_006722582.2 | XP_006722645.1 | 10 | 1743 | ||
XM_011526268.2 | XP_011524570.1 | 12 | 1934 | ||
GenBank mRNA | AF038960.1 | - | 11 | 1796 | |
AF195514.1 | - | 11 | 1808 | ||
BC039574.1 | - | 11 | 1774 | 6042862 |