Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_010859.2 | NP_034989.1 | 7 | 716 | |
GenBank mRNA | AK002312.1 | - | 7 | 718 | |
AK011518.1 | - | 7 | 709 | ||
AK146943.1 | - | 7 | 720 | ||
BC061222.1 | - | 7 | 719 | 30297023 | |
X67685.1 | - | 4 | 216 |