Gene Transcripts

Gene Symbol
Ncf2
Entrez Gene ID
17970
Gene Name
neutrophil cytosolic factor 2
Gene Aliases
NOXA2, Ncf-2, p67phox
Chromosome Location
Chr. 1: 152800158 - 152837169 on Build GRCm38
UniGene ID
Mm.270307
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010877.4 NP_035007.1 2 382
GenBank mRNA AB002664.1 - 2 336
AK036379.1 - 2 383
AK137152.1 - 2 437
AK153447.1 - 2 344
AK155324.1 - 2 437
AK170839.1 - 2 407
AK170998.1 - 2 410
AK171059.1 - 2 408
AK171265.1 - 2 410
BC003730.1 - 2 330 3592460
BC076609.1 - 3 1336
BC094056.1 - 2 428
DQ449939.1 - 2 337

View Full Product Details