Gene Transcripts

Gene Symbol
Oit3
Entrez Gene ID
18302
Gene Name
oncoprotein induced transcript 3
Gene Aliases
EF-9, LZP
Chromosome Location
Chr. 10: 59422960 - 59441779 on Build GRCm38
UniGene ID
Mm.123013
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010959.2 NP_035089.1 9 2237
GenBank mRNA AF517107.1 - 8 2056
AK040570.1 - 9 2238
AK153440.1 - 5 1479
AY180915.1 - 9 2161

View Full Product Details