Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001083316.1 | NP_001076785.1 | 4 | 806 | |
NM_011058.2 | NP_035188.2 | 4 | 789 | ||
GenBank mRNA | AK035501.1 | - | 4 | 746 | |
AK081664.1 | - | 4 | 788 | ||
AK136490.1 | - | 4 | 775 | ||
AK139094.1 | - | 3 | 1021 | ||
AK145097.1 | - | 4 | 1389 | ||
AK147267.1 | - | 4 | 738 | ||
AK163952.1 | - | 4 | 853 | ||
BC020405.1 | - | 4 | 720 | ||
BC037467.1 | - | 4 | 728 | ||
BC053036.1 | - | 4 | 806 | 5704645 | |
DQ912857.1 | - | 1 | 133 | ||
M84607.1 | - | 4 | 799 |