Gene Transcripts

Gene Symbol
ZMYM4
Entrez Gene ID
9202
Gene Name
zinc finger MYM-type containing 4
Gene Aliases
CDIR, MYM, ZNF198L3, ZNF262
Chromosome Location
Chr. 1: 35268694 - 35421944 on Build GRCh38
UniGene ID
Hs.269211
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005095.2 NP_005086.2 7 1158
XM_005271328.4 XP_005271385.1 7 1437
XM_005271331.2 XP_005271388.1 4 403
XM_011542421.2 XP_011540723.1 7 1084
XM_011542423.2 XP_011540725.1 8 1489
XM_011542425.2 XP_011540727.1 5 443
XM_017002803.1 XP_016858292.1 8 1211
XM_017002804.1 XP_016858293.1 8 1151
GenBank mRNA AB007885.1 - 3 278
AK308320.1 - 7 1159
AK309705.1 - 6 818
BC012093.1 - 5 737
BC127113.1 - 8 1164 40125110
BC127114.1 - 4 347 40125115
BX647376.1 - 3 388
CR936741.1 - 6 1022

View Full Product Details