Gene Transcripts

Gene Symbol
Fpr2
Entrez Gene ID
14289
Gene Name
formyl peptide receptor 2
Gene Aliases
E330010I07Rik, Fpr-rs2
Chromosome Location
Chr. 17: 17887824 - 17893952 on Build GRCm38
UniGene ID
Mm.378918
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008039.2 NP_032065.1 2 1102
GenBank mRNA AK054286.1 - 3 691
BC117066.1 - 1 1038 40126385

View Full Product Details