Gene Transcripts

Gene Symbol
Gif
Entrez Gene ID
14603
Gene Name
gastric intrinsic factor
Gene Aliases
AV073125
Chromosome Location
Chr. 19: 11747543 - 11763447 on Build GRCm38
UniGene ID
Mm.456
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008118.3 NP_032144.2 4 589
GenBank mRNA AK078933.1 - 4 590
BC118519.1 - 4 575 40105765
L24191.1 - 4 501

View Full Product Details