Gene Transcripts
- Gene Symbol
- Hsd17b4
- Entrez Gene ID
- 15488
- Gene Name
- hydroxysteroid (17-beta) dehydrogenase 4
- Gene Aliases
- 17-beta-HSD, 17[b]-HSD, 4, DBP, MFE-2, MFP2, MPF-2, Mfp-2, perMFE-2
- Chromosome Location
- Chr. 18: 50128201 - 50196270 on Build GRCm38
- UniGene ID
- Mm.277857
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC022175.1 |
- |
2, 3 |
132 |
|
View Full Product Details