Gene Transcripts

Gene Symbol
Il7r
Entrez Gene ID
16197
Gene Name
interleukin 7 receptor
Gene Aliases
CD127, IL-7Ralpha
Chromosome Location
Chr. 15: 9506159 - 9529876 on Build GRCm38
UniGene ID
Mm.389
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008372.4 NP_032398.3 8 2908
GenBank mRNA AA183527.1 - 1 165
BC089571.1 - 8 2843 6758487

View Full Product Details