Gene Transcripts

Gene Symbol
ISG15
Entrez Gene ID
9636
Gene Name
ISG15 ubiquitin-like modifier
Gene Aliases
G1P2, IFI15, IMD38, IP17, UCRP, hUCRP
Chromosome Location
Chr. 1: 1013467 - 1014540 on Build GRCh38
UniGene ID
Hs.458485
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_005101.3 NP_005092.1 2 644
GenBank mRNA BC009507.2 - 2 578
BU161226.1 - 2 601
M13755.1 - 2 612

View Full Product Details