Gene Transcripts
- Gene Symbol
- Frk
- Entrez Gene ID
- 14302
- Gene Name
- fyn-related kinase
- Gene Aliases
- BSK, BSK/IYK, C85044, GTK, RAK
- Chromosome Location
- Chr. 10: 34483400 - 34611534 on Build GRCm38
- UniGene ID
- Mm.332432
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001159544.1 |
NP_001153016.1
|
1, 2 |
976 |
|
NM_010237.3 |
NP_034367.2
|
2, 3 |
820 |
|
GenBank mRNA |
AK028855.1 |
- |
2, 3 |
760 |
|
AK052614.1 |
- |
2, 3 |
692 |
|
AK077264.1 |
- |
1, 2 |
845 |
|
AK085191.1 |
- |
1, 2 |
821 |
|
AK166379.1 |
- |
2, 3 |
820 |
|
BC007137.1 |
- |
2, 3 |
833 |
3601246 |
L36132.1 |
- |
1, 2 |
855 |
|
Z48757.1 |
- |
2, 3 |
685 |
|
View Full Product Details