Gene Transcripts

Gene Symbol
Ids
Entrez Gene ID
15931
Gene Name
iduronate 2-sulfatase
Gene Aliases
AW214631
Chromosome Location
Chr. X: 70343069 - 70365114 on Build GRCm38
UniGene ID
Mm.233083
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010498.3 NP_034628.2 5 776
GenBank mRNA AJ627044.1 - 3 241
AK085237.1 - 6 653
AK132287.1 - 6 656
AK166178.1 - 5 654
AK166443.1 - 6 605
L07921.1 - 5 653

View Full Product Details