Gene Transcripts

Gene Symbol
Il18bp
Entrez Gene ID
16068
Gene Name
interleukin 18 binding protein
Gene Aliases
IL-18BP, Igifbp, MC54L
Chromosome Location
Chr. 7: 102015077 - 102019041 on Build GRCm38
UniGene ID
Mm.422756
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_010531.1 NP_034661.1 6 569
GenBank mRNA AB019505.1 - 4 418
AF110802.1 - 4 1796
AF110803.1 - 5 496
AF122907.1 - 6 569
AK003370.1 - 6 585
AK008452.1 - 5 500
AK009721.1 - 5 496
AK009877.1 - 2 136
BC018332.1 - 5 479 4007411
CT010164.1 - 4 418
CT010334.1 - 4 418

View Full Product Details