Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013536.2 | NP_038564.1 | 6 | 802 | |
GenBank mRNA | AA185226.1 | - | 2 | 132 | |
AK081757.1 | - | 6 | 780 | ||
AK178640.1 | - | 2 | 182 | ||
AK218607.1 | - | 4 | 323 | ||
BC002004.1 | - | 6 | 722 | 3487480 |