Gene Transcripts

Gene Symbol
H2-Ke6
Entrez Gene ID
14979
Gene Name
H2-K region expressed gene 6
Gene Aliases
D17H6S112E, H-2Ke6, Hsd17b8, Ke-6, Ke6, Ring2
Chromosome Location
Chr. 17: 34026033 - 34028055 on Build GRCm38
UniGene ID
Mm.275452
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013543.2 NP_038571.2 5, 6 568
GenBank mRNA BC086927.1 - 5, 6 568
CX243612.1 - 5, 6 537
KJ650217.1 - 5, 6 560

View Full Product Details