Gene Transcripts
- Gene Symbol
- Hnrnpk
- Entrez Gene ID
- 15387
- Gene Name
- heterogeneous nuclear ribonucleoprotein K
- Gene Aliases
- Hnrpk, KBBP, NOVA
- Chromosome Location
- Chr. 13: 58391132 - 58403343 on Build GRCm38
- UniGene ID
- Mm.142872
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001301340.1 |
NP_001288269.1
|
5, 6 |
410 |
|
NM_001301341.1 |
NP_001288270.1
|
7, 8 |
537 |
|
NM_001301343.1 |
NP_001288272.1
|
5, 6 |
396 |
|
NM_001301344.1 |
NP_001288273.1
|
5, 6 |
410 |
|
NM_001301345.1 |
NP_001288274.1
|
5, 6 |
410 |
|
NM_025279.3 |
NP_079555.1
|
5, 6 |
410 |
|
GenBank mRNA |
AK011428.1 |
- |
5, 6 |
415 |
|
AK051313.1 |
- |
5, 6 |
404 |
|
AK078777.1 |
- |
7, 8 |
538 |
|
AK088462.1 |
- |
5, 6 |
397 |
|
AK128891.1 |
- |
5, 6 |
403 |
|
AK135519.1 |
- |
5, 6 |
324 |
|
AK144245.1 |
- |
|
7620 |
|
AK150848.1 |
- |
5, 6 |
390 |
|
AK151689.1 |
- |
5, 6 |
364 |
|
AK152927.1 |
- |
5, 6 |
364 |
|
AK160615.1 |
- |
5, 6 |
380 |
|
AK160887.1 |
- |
5, 6 |
412 |
|
AK166656.1 |
- |
5, 6 |
415 |
|
AK166927.1 |
- |
5, 6 |
368 |
|
AK167600.1 |
- |
5, 6 |
401 |
|
AK168918.1 |
- |
1, 2 |
313 |
|
AK169064.1 |
- |
5, 6 |
400 |
|
BC006694.1 |
- |
5, 6 |
377 |
3583037 |
BC089328.1 |
- |
5, 6 |
333 |
6402614 |
EU604547.1 |
- |
4, 5 |
263 |
|
View Full Product Details