Gene Transcripts
- Gene Symbol
- Gpx2
- Entrez Gene ID
- 14776
- Gene Name
- glutathione peroxidase 2
- Gene Aliases
- GI-GPx, GPx-GI, GSHPx-2, GSHPx-GI
- Chromosome Location
- Chr. 12: 76792335 - 76795554 on Build GRCm38
- UniGene ID
- Mm.441856
- Species
- Mus musculus
- Gene Symbol
- Gpx2-ps1
- Entrez Gene ID
- 14777
- Gene Name
- glutathione peroxidase 2, pseudogene 1
- Gene Aliases
- AU041318
- Chromosome Location
- Chr. 7: 100264544 - 100265561 on Build GRCm38
- UniGene ID
- Mm.57225
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BG084322.2 |
- |
1 |
422 |
|
View Full Product Details