Gene Transcripts

Gene Symbol
Ap2a1
Entrez Gene ID
11771
Gene Name
adaptor-related protein complex 2, alpha 1 subunit
Gene Aliases
Adtaa
Chromosome Location
Chr. 7: 44900373 - 44929490 on Build GRCm38
UniGene ID
Mm.6877
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077264.1 NP_001070732.1 5 723
NM_007458.2 NP_031484.1 5 723
GenBank mRNA AK135424.1 - 5 652
AK155515.1 - 5 659
BC031433.1 - 5 673 4481643
CA327188.1 - 5 729

View Full Product Details