Gene Transcripts

Gene Symbol
Bmp8a
Entrez Gene ID
12163
Gene Name
bone morphogenetic protein 8a
Gene Aliases
Bmp7r1, OP-2, OP2
Chromosome Location
Chr. 4: 123312645 - 123343252 on Build GRCm38
UniGene ID
Mm.439749
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001256019.1 NP_001242948.1 7 1755
NM_007558.3 NP_031584.1 7 1716
GenBank mRNA AK157978.1 - 7 1378
BC052168.1 - 7 1753 6334507
M97017.1 - 7 1264

View Full Product Details