Gene Transcripts

Gene Symbol
Cdr2
Entrez Gene ID
12585
Gene Name
cerebellar degeneration-related 2
Gene Aliases
AA617262
Chromosome Location
Chr. 7: 120957036 - 120982312 on Build GRCm38
UniGene ID
Mm.1640
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007672.2 NP_031698.2 2 439
GenBank mRNA AK148057.1 - 2 423
AK171706.1 - 2 394
BC117530.1 - 2 328 40106297
BC118511.1 - 2 328 40106298
BY713743.1 - 2 439
U88588.1 - 3 332

View Full Product Details