Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007751.3 | NP_031777.1 | 2 | 320 | |
GenBank mRNA | AA028501.1 | - | 2 | 297 | |
AA511205.1 | - | 2 | 274 | ||
AK003116.1 | - | 2 | 263 | ||
BC027531.1 | - | 2 | 264 | 1515187 | |
BC086930.1 | - | 2 | 283 | 6752860 | |
BE626379.1 | - | 2 | 320 | ||
U15541.1 | - | 2 | 270 |