Gene Transcripts

Gene Symbol
Ctsk
Entrez Gene ID
13038
Gene Name
cathepsin K
Gene Aliases
AI323530, MMS10-Q, Ms10q, catK
Chromosome Location
Chr. 3: 95499210 - 95509387 on Build GRCm38
UniGene ID
Mm.272085
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007802.4 NP_031828.2 8 1052
GenBank mRNA AK003425.1 - 8 977
AK132648.1 - 8 982
BC046320.1 - 8 1006 6390854
X94444.1 - 7 924

View Full Product Details