Gene Transcripts
- Gene Symbol
- Amy2a5
- Entrez Gene ID
- 109959
- Gene Name
- amylase 2a5
- Gene Aliases
- 1810008N23Rik, Amy-2, Amy2, mAmy2-2
- Chromosome Location
- Chr. 3: 113249178 - 113258699 on Build GRCm38
- UniGene ID
- Mm.275426
- Species
- Mus musculus
- Gene Symbol
- Amy2a2
- Entrez Gene ID
- 100043688
- Gene Name
- amylase 2a2
- Gene Aliases
- ENSMUSG00000074265, OTTMUSG00000022431, mAmy2-5
- Chromosome Location
- Chr. 3: 113345049 - 113356658 on Build GRCm38
- UniGene ID
- Mm.275426
- Species
- Mus musculus
- Gene Symbol
- Amy2a3
- Entrez Gene ID
- 100043686
- Gene Name
- amylase 2a3
- Gene Aliases
- OTTMUSG00000022430, mAmy2-4
- Chromosome Location
- Chr. 3: 113312449 - 113324052 on Build GRCm38
- UniGene ID
- Mm.275426
- Species
- Mus musculus
- Gene Symbol
- Amy2a4
- Entrez Gene ID
- 100043684
- Gene Name
- amylase 2a4
- Gene Aliases
- ENSMUSG00000074267, OTTMUSG00000022433, mAmy2-3
- Chromosome Location
- Chr. 3: 113279837 - 113291449 on Build GRCm38
- UniGene ID
- Mm.275426
- Species
- Mus musculus
- Gene Symbol
- Amy2b
- Entrez Gene ID
- 545562
- Gene Name
- amylase 2b
- Gene Aliases
- Amy-X, Amy2-2, OTTMUSG00000022462, mAmy2-1
- Chromosome Location
- Chr. 3: 113147656 - 113156727 on Build GRCm38
- UniGene ID
- Mm.462977
- Species
- Mus musculus
View Full Product Details