Gene Transcripts
- Gene Symbol
- UBE2K
- Entrez Gene ID
- 3093
- Gene Name
- ubiquitin conjugating enzyme E2 K
- Gene Aliases
- E2-25K, HIP2, HYPG, LIG, UBC1
- Chromosome Location
- Chr. 4: 39698044 - 39782792 on Build GRCh38
- UniGene ID
- Hs.50308
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001111112.1 |
NP_001104582.1
|
2, 3 |
449 |
|
NM_001312646.1 |
NP_001299575.1
|
2, 3 |
449 |
|
NM_001312647.1 |
NP_001299576.1
|
2, 3 |
361 |
|
NM_005339.4 |
NP_005330.1
|
2, 3 |
449 |
|
GenBank mRNA |
AB022435.1 |
- |
2, 3 |
251 |
|
AK093533.1 |
- |
2, 3 |
270 |
|
AK295846.1 |
- |
2, 3 |
384 |
|
AK315524.1 |
- |
2, 3 |
375 |
|
BC022804.1 |
- |
2, 3 |
325 |
4893778 |
BC050600.1 |
- |
2, 3 |
303 |
6502901 |
CX786829.1 |
- |
2, 3 |
288 |
|
DA526283.1 |
- |
2, 3 |
449 |
|
U58522.1 |
- |
2, 3 |
168 |
|
View Full Product Details