Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001111058.1 | NP_001104528.1 | 7 | 1086 | |
NM_021293.3 | NP_067268.1 | 6 | 1003 | ||
GenBank mRNA | AK046303.1 | - | 8 | 1226 | |
AK155745.1 | - | 6 | 933 | ||
AK170937.1 | - | 6 | 971 | ||
BC132379.1 | - | 6 | 1003 | 40130656 | |
S71345.1 | - | 7 | 1017 | ||
S71403.1 | - | 6 | 934 |