Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133653.3 | NP_598414.1 | 10 | 1659 | |
GenBank mRNA | AK149542.1 | - | 9 | 1410 | |
AK153832.1 | - | 9 | 1413 | ||
BC011211.1 | - | 9 | 1397 | 4196928 | |
L13622.1 | - | 9 | 1411 |