Gene Transcripts

Gene Symbol
Adam7
Entrez Gene ID
11500
Gene Name
a disintegrin and metallopeptidase domain 7
Gene Aliases
9230104J12Rik, EAP1, EAPI
Chromosome Location
Chr. 14: 68497336 - 68533689 on Build GRCm38
UniGene ID
Mm.261495
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007402.2 NP_031428.2 18 1979
GenBank mRNA AF013107.1 - 18 1965
AK136692.1 - 18 1978
AK164965.1 - 4 499

View Full Product Details