Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009627.1 | NP_033757.1 | 4 | 1014 | |
GenBank mRNA | AK144181.1 | - | 4 | 1029 | |
AK144600.1 | - | 4 | 1023 | ||
AK144649.1 | - | 4 | 1027 | ||
BC052665.1 | - | 4 | 1024 | 30054008 | |
U77630.1 | - | 4 | 1014 |