Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001083119.2 | NP_001076588.1 | 12 | 2230 | |
NM_011214.2 | NP_035344.2 | 12 | 2230 | ||
GenBank mRNA | AK047640.1 | - | 11 | 1928 | |
AK052720.1 | - | 12 | 2205 | ||
BC080736.1 | - | 12 | 2185 | ||
D88187.1 | - | 12 | 2221 |