Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001272035.1 | NP_001258964.1 | 5 | 1834 | |
NM_015728.5 | NP_056543.2 | 5 | 1834 | ||
GenBank mRNA | AB016795.1 | - | 5 | 1291 | |
AK144703.1 | - | 5 | 1701 | ||
AW519598.1 | - | 3 | 313 | ||
BC005616.1 | - | 5 | 1411 | 3154886 | |
BC054395.1 | - | 5 | 1834 | 6492236 | |
BC063104.1 | - | 1767 | 5042025 |