Gene Transcripts

Gene Symbol
Tusc2
Entrez Gene ID
80385
Gene Name
tumor suppressor candidate 2
Gene Aliases
1190001E22Rik, AA407686, Fus1, Lgcc, PAP, Pdap2
Chromosome Location
Chr. 9: 107563255 - 107566108 on Build GRCm38
UniGene ID
Mm.21739
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019742.4 NP_062716.1 3 740
GenBank mRNA AF123387.1 - 3 627
AK082684.1 - 3 741

View Full Product Details