Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023162.4 | NP_075651.1 | 2 | 369 | |
GenBank mRNA | AF230339.1 | - | 2 | 242 | |
AK003714.1 | - | 2 | 370 | ||
AK005976.1 | - | 3 | 342 | ||
AK019152.1 | - | 3 | 346 | ||
BC043016.1 | - | 3 | 306 | 5683859 |