Gene Transcripts

Gene Symbol
CAMK4
Entrez Gene ID
814
Gene Name
calcium/calmodulin dependent protein kinase IV
Gene Aliases
CaMK IV, CaMK-GR, caMK
Chromosome Location
Chr. 5: 111223653 - 111498503 on Build GRCh38
UniGene ID
Hs.438801
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001323374.1 NP_001310303.1 5 474
NM_001323375.1 NP_001310304.1 5 506
NM_001744.5 NP_001735.1 4 622
GenBank mRNA AK312840.1 - 4 477
BC016695.1 - 4 466
BC025687.1 - 4 464 5205478
D30742.1 - 4 452
L17000.1 - 4 416
L24959.1 - 5 474

View Full Product Details