Gene Transcripts

Gene Symbol
Ahctf1
Entrez Gene ID
226747
Gene Name
AT hook containing transcription factor 1
Gene Aliases
6230412P20Rik, AV011447, Elys
Chromosome Location
Chr. 1: 179744894 - 179804015 on Build GRCm38
UniGene ID
Mm.128165
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026375.2 NP_080651.2 3 867
GenBank mRNA AB059278.1 - 3 523
BC138343.1 - 3 510 8861363
BC138344.1 - 3 510 8861364

View Full Product Details