Gene Transcripts

Gene Symbol
Krtcap3
Entrez Gene ID
69815
Gene Name
keratinocyte associated protein 3
Gene Aliases
2010001C09Rik, AW321088, Kcp3
Chromosome Location
Chr. 5: 31251706 - 31253197 on Build GRCm38
UniGene ID
Mm.389609
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027221.3 NP_081497.2 7 868
GenBank mRNA AK007999.1 - 7 785
BC016516.1 - 6 982 2609442
BC027812.1 - 7 866 5251276

View Full Product Details