Gene Transcripts
- Gene Symbol
- Acsl6
- Entrez Gene ID
- 216739
- Gene Name
- acyl-CoA synthetase long-chain family member 6
- Gene Aliases
- A330035H04Rik, AW050338, Facl6, LACS, Lacsl, mKIAA0837
- Chromosome Location
- Chr. 11: 54303824 - 54364756 on Build GRCm38
- UniGene ID
- Mm.267478
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001033597.1 |
NP_001028769.1
|
16, 17 |
1680 |
|
NM_001033598.1 |
NP_001028770.1
|
16, 17 |
1649 |
|
NM_001033599.1 |
NP_001028771.1
|
16, 17 |
1855 |
|
NM_144823.4 |
NP_659072.3
|
16, 17 |
1680 |
|
GenBank mRNA |
AB073974.1 |
- |
15, 16 |
1524 |
|
AK122384.1 |
- |
5, 6 |
3446 |
|
AK147283.1 |
- |
16, 17 |
1650 |
|
AK147421.1 |
- |
16, 17 |
1681 |
|
AY167035.1 |
- |
16, 17 |
1650 |
|
AY786360.1 |
- |
18, 19 |
2198 |
|
AY786361.1 |
- |
16, 17 |
1630 |
|
AY786362.1 |
- |
16, 17 |
1822 |
|
AY786363.1 |
- |
16, 17 |
1675 |
|
BC016114.1 |
- |
16, 17 |
1606 |
4481949 |
BC022959.1 |
- |
16, 17 |
1680 |
5362660 |
View Full Product Details