Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_144851.5 | NP_659100.1 | 4 | 488 | |
GenBank mRNA | AK028565.1 | - | 4 | 230 | |
AK035581.1 | - | 4 | 324 | ||
AK035723.1 | - | 4 | 592 | ||
AK054521.1 | - | 4 | 489 | ||
AK088597.1 | - | 4 | 232 | ||
AK138013.1 | - | 4 | 289 | ||
BC023129.1 | - | 3 | 202 | 3595534 |