Gene Transcripts

Gene Symbol
TROAP
Entrez Gene ID
10024
Gene Name
trophinin associated protein
Gene Aliases
TASTIN
Chromosome Location
Chr. 12: 49322135 - 49331733 on Build GRCh38
UniGene ID
Hs.524399
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100620.1 NP_001094090.1 2, 3 318
NM_001278324.1 NP_001265253.1 2, 3 318
NM_005480.3 NP_005471.3 2, 3 318
XM_006719181.1 XP_006719244.1 2, 3 262
XM_011537723.1 XP_011536025.1 2, 3 262
XM_011537725.2 XP_011536027.1 3, 4 454
GenBank mRNA AK296811.1 - 2, 3 251
AK299302.1 - 2, 3 223
AK315211.1 - 2, 3 262
BC011597.2 - 2, 3 186
BC032583.1 - 2, 3 230 5498800
DC410015.1 - 2, 3 318
U04810.2 - 2, 3 261

View Full Product Details