Gene Transcripts

Gene Symbol
Vmn2r81
Entrez Gene ID
216144
Gene Name
vomeronasal 2, receptor 81
Gene Aliases
EC1-V2R, EC1-VR2, V2rf2
Chromosome Location
Chr. 10: 79247777 - 79294535 on Build GRCm38
UniGene ID
Mm.246574
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175936.1 NP_787950.1 3 732
GenBank mRNA AJ543404.1 - 3 732

View Full Product Details