Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_011327.4 | NP_035457.1 | 3 | 243 | |
GenBank mRNA | AK004860.2 | - | 3 | 173 | |
BC018384.1 | - | 3 | 198 | 3710340 | |
BC034613.1 | - | 3 | 164 | 5123611 | |
DN176201.1 | - | 3 | 243 | ||
M91458.1 | - | 3 | 158 |