Gene Transcripts

Gene Symbol
Rhot1
Entrez Gene ID
59040
Gene Name
ras homolog gene family, member T1
Gene Aliases
2210403N23Rik, AA415293, AF244542, AI834919, Arht1, C430039G08Rik, Miro1
Chromosome Location
Chr. 11: 80209005 - 80267907 on Build GRCm38
UniGene ID
Mm.489674
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163354.1 NP_001156826.1 9 801
NM_001163355.1 NP_001156827.1 9 801
NM_021536.7 NP_067511.4 9 801
GenBank mRNA AK019059.1 - 9 761
AK028597.1 - 9 744
AK031139.1 - 7 463
AK032096.1 - 9 774
AK041119.1 - 9 737
AK053263.1 - 8 658
AK054472.1 - 9 772
AK078955.1 - 9 744
AK090071.1 - 2 127
AK138830.1 - 9 742
AK141621.1 - 9 628
AK147749.1 - 9 780
AK147801.1 - 9 806
AK167502.1 - 9 801
AK169811.1 - 9 770
BC046785.1 - 9 784 6416127
BC058350.1 - 9 716 6815832

View Full Product Details