Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028994.2 | NP_083270.1 | 8 | 1469 | |
GenBank mRNA | AK007429.1 | - | 5 | 662 | |
AK030501.1 | - | 8 | 1470 | ||
AK031612.1 | - | 8 | 1314 | ||
AK034927.1 | - | 8 | 1348 | ||
AK142561.1 | - | 2 | 221 | ||
AK147972.1 | - | 8 | 1323 | ||
AK165094.1 | - | 8 | 1347 | ||
AK170242.1 | - | 8 | 1323 | ||
AK218043.1 | - | 2 | 64 | ||
AK219654.1 | - | 2 | 227 | ||
BC010318.1 | - | 8 | 1332 | 3596227 | |
BC011126.1 | - | 7 | 1631 | ||
BC023437.1 | - | 6 | 937 | ||
BC028326.1 | - | 3 | 270 |