Gene Transcripts

Gene Symbol
Rhot2
Entrez Gene ID
214952
Gene Name
ras homolog gene family, member T2
Gene Aliases
Arht2, Miro2
Chromosome Location
Chr. 17: 25838838 - 25844851 on Build GRCm38
UniGene ID
Mm.40907
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145999.2 NP_666111.1 19 2775
GenBank mRNA AY311390.1 - 19 2775
BC023163.1 - 9 1544
BC023927.1 - 5 1828
BC029777.1 - 19 2408 4950061
BC034062.1 - 19 2430 4207654

View Full Product Details