Gene Transcripts

Gene Symbol
Zdhhc17
Entrez Gene ID
320150
Gene Name
zinc finger, DHHC domain containing 17
Gene Aliases
A230053P19Rik, BB187739, D130071N24Rik, Hip14
Chromosome Location
Chr. 10: 110941779 - 111010066 on Build GRCm38
UniGene ID
Mm.339281
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172554.2 NP_766142.2 2 180
GenBank mRNA AK038670.1 - 2 124
AK038681.1 - 2 124
AK039575.1 - 2 181
AK046891.1 - 2 122
AK122406.1 - 2 254
AK134967.1 - 2 129
AK162922.1 - 2 118
BC027101.1 - 2 109
BC058772.1 - 2 121
BC080664.1 - 2 119
BC117788.1 - 2 122 40092137
BC117789.2 - 2 122 40092140

View Full Product Details