Gene Transcripts

Gene Symbol
Zdhhc17
Entrez Gene ID
320150
Gene Name
zinc finger, DHHC domain containing 17
Gene Aliases
A230053P19Rik, BB187739, D130071N24Rik, Hip14
Chromosome Location
Chr. 10: 110941779 - 111010066 on Build GRCm38
UniGene ID
Mm.339281
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172554.2 NP_766142.2 16 1763
GenBank mRNA AK039575.1 - 16 1764
AK046891.1 - 16 1705
AK122406.1 - 15 2708
BC027101.1 - 16 1651
BC117788.1 - 16 1705 40092137
BC117789.2 - 16 1705 40092140

View Full Product Details