Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_172653.2 | NP_766241.2 | 10 | 2967 | |
GenBank mRNA | AK030685.1 | - | 1102 | ||
AK049099.1 | - | 8 | 1713 | ||
AK122483.1 | - | 9 | 2401 | ||
BC059214.1 | - | 8 | 1851 | ||
BC062918.1 | - | 10 | 2967 | 6833098 |